Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 3-mercaptopropionate 98%, ACROS Organics™

Click to view available options
Quantity:
5g
100g
500g
Packaging:
Glass bottle
2935-90-2
C4H8O2S
120.17
MFCD00004895
LDTLDBDUBGAEDT-UHFFFAOYSA-N
methyl 3-mercaptopropionate,propanoic acid, 3-mercapto-, methyl ester,3-mercaptopropionic acid methyl ester,methyl mercaptopropionate,methyl3-mercaptopropionate,methyl 3-mercaptopropanoate,propionic acid, 3-mercapto-, methyl ester,unii-v920f0bcrz,methyl-3-mercaptopropionate,v920f0bcrz
18050
methyl 3-sulfanylpropanoate
COC(=O)CCS
2935-90-2
C4H8O2S
120.17
MFCD00004895
LDTLDBDUBGAEDT-UHFFFAOYSA-N
methyl 3-mercaptopropionate,propanoic acid, 3-mercapto-, methyl ester,3-mercaptopropionic acid methyl ester,methyl mercaptopropionate,methyl3-mercaptopropionate,methyl 3-mercaptopropanoate,propionic acid, 3-mercapto-, methyl ester,unii-v920f0bcrz,methyl-3-mercaptopropionate,v920f0bcrz
18050
methyl 3-sulfanylpropanoate
COC(=O)CCS
Density 1.0800g/mL
Packaging Glass bottle
Specific Gravity 1.08
Boiling Point 166.0°C to 169.0°C
Infrared Spectrum Authentic
Flash Point 60°C
Refractive Index 1.4620 to 1.4640
Quantity 5g
Color Colorless
Assay Percent Range 98%
Linear Formula HSCH2CH2CO2CH3
Beilstein 03, II, 214
Formula Weight 120.17
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material Methyl 3-mercaptopropionate, 98%
Show More Show Less

GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Flammable liquid and vapor.
Toxic if swallowed.
Harmful in contact with skin.
Fatal if inhaled.
Very toxic to aquatic life with long lasting effects.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

GHS Signal Word: Danger

EINECSNumber : 220-912-4

TSCA : TSCA