Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 3-(methylthio)propionate, 99+%, Thermo Scientific™

Catalog No. p-165317
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13532-18-8
Molecular Formula C5H10O2S
Molecular Weight (g/mol) 134.19
MDL Number MFCD00008565
InChI Key DMMJVMYCBULSIS-UHFFFAOYSA-N
Synonym methyl 3-methylthio propionate, methyl 3-methylthiopropionate, methyl 3-methylthio propanoate, propanoic acid, 3-methylthio-, methyl ester, methyl beta-methiopropionate, methyl 3-methylsulfanyl propanoate, methyl-3-methylthio propionate, methyl 3-methylthiopropanoate, propionic acid, 3-methylthio-, methyl ester, methyl beta-methylthiopropionate
PubChem CID 61641
IUPAC Name methyl 3-methylsulfanylpropanoate
SMILES COC(=O)CCSC

Specifications

Color Colorless
Density 1.0700g/mL
Boiling Point 74.0°C to 75.0°C (13.0 mmHg)
Flash Point 72°C
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Refractive Index 1.4640 to 1.4660
Linear Formula CH3SCH2CH2CO2CH3
Quantity 5 g
Specific Gravity 1.07
Solubility Information Solubility in water: insoluble.
Formula Weight 134.19
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material Methyl 3-(methylthio)propionate, 99+%
Show More Show Less
Hazard Category STENCH
EINECSNumber 236-883-6
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.