Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Methyl 3-nitrobenzoate, 98+%
Click to view available options
Quantity:
50 g
1000 g
Description
Methyl 3-nitrobenzoate was used in the preparation of iodoarene.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Methyl 3-nitrobenzoate was used in the preparation of iodoarene.
Solubility
Insoluble in water.
Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
Methyl 3-nitrobenzoate was used in the preparation of iodoarene.
Solubility
Insoluble in water.
Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.

Specifications
Specifications
Melting Point | 77°C to 80°C |
Boiling Point | 279°C |
Quantity | 50 g |
Beilstein | 392449 |
Solubility Information | Insoluble in water. |
Formula Weight | 181.15 |
Percent Purity | ≥98% |
Chemical Name or Material | Methyl 3-nitrobenzoate |
Safety and Handling
EINECSNumber | 210-573-0 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction