Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 4-amino-3-chlorobenzoate, 97%, Thermo Scientific Chemicals

Catalog No. p-159937
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 84228-44-4
Molecular Formula C8H8ClNO2
Molecular Weight (g/mol) 185.61
MDL Number MFCD01320686
InChI Key WKVJBYKFGNVWLM-UHFFFAOYSA-N
Synonym 4-amino-3-chlorobenzoic acid methyl ester, 4-amino-3-chloro-benzoic acid methyl ester, methyl 4-amino-3-chloro-benzoate, benzoic acid, 4-amino-3-chloro-, methyl ester, 2-chloro-4-methoxycarbonyl aniline, pubchem10804, akos bb-3098, timtec-bb sbb003712, acmc-20a51m, methyl-4-amino-3-chlorobenzoate
PubChem CID 1515284
IUPAC Name methyl 4-amino-3-chlorobenzoate
SMILES COC(=O)C1=CC(=C(C=C1)N)Cl

Specifications

Melting Point 106°C to 110°C
Color Beige to Brown
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Linear Formula NH2C6H3(Cl)COOH
Quantity 1 g
Formula Weight 185.61
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material Methyl 4-amino-3-chlorobenzoate, 97%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breath

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.