Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 4-Amino-3-methylbenzoate 98.0+%, TCI America™
SDP

Catalog No. M215225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M215225G 25 g
M21525G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M215225G Supplier TCI America Supplier No. M215225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 18595-14-7
Molecular Formula C9H11NO2
Molecular Weight (g/mol) 165.19
MDL Number MFCD00102230
InChI Key ZHIPSMIKSRYZFV-UHFFFAOYSA-N
Synonym 4-amino-3-methylbenzoic acid methyl ester, 4-amino-3-toluic acid methyl ester, methyl 3-methyl-4-aminobenzoate, methyl-4-amino-3-methyl-benzoate, methyl 4-amino-3-methyl-benzoate, benzoic acid, 4-amino-3-methyl-, methyl ester, 4-amino-m-toluic acid methyl ester, methyl 4-amino-3-methylbenzenecarboxylate, zlchem 944, methyl 4-amino-m-toluate
PubChem CID 2736799
IUPAC Name methyl 4-amino-3-methylbenzoate
SMILES COC(=O)C1=CC=C(N)C(C)=C1

Specifications

Melting Point 120°C
Quantity 25 g
Formula Weight 165.19
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 4-Amino-3-methylbenzoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.