Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 4-aminobenzoate, 98%

Catalog No. AAA1171514
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1171514 25 g
AAA1171522 100 g
AAA1171536 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1171514 Supplier Thermo Scientific Chemicals Supplier No. A1171514
Only null left
Add to Cart

CAS: 619-45-4 | C8H9NO2 | 151.165 g/mol

Methyl 4-aminobenzoate is used in the synthesis of guanidine alkaloids, (±)-martinelline and (±)-martinellic acid by a protic acid catalyzed hetero Diels-Alder coupling reaction with N-Cbz 2-pyrroline. It is also used in organic synthesis and acts as a dye intermediate. Further, it is used to prepare 4-(2-methyl-4-oxo-4H-quinazolin-3-yl)-benzoic acid methyl ester by reacting with 2-acetylamino-benzoic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 4-aminobenzoate is used in the synthesis of guanidine alkaloids, (±)-martinelline and (±)-martinellic acid by a protic acid catalyzed hetero Diels-Alder coupling reaction with N-Cbz 2-pyrroline. It is also used in organic synthesis and acts as a dye intermediate. Further, it is used to prepare 4-(2-methyl-4-oxo-4H-quinazolin-3-yl)-benzoic acid methyl ester by reacting with 2-acetylamino-benzoic acid.

Solubility
Soluble in alcohol and ether. Slightly soluble in water.

Notes
Incompatible with strong oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 619-45-4
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.165
MDL Number MFCD00007891
InChI Key LZXXNPOYQCLXRS-UHFFFAOYSA-N
Synonym 4-aminobenzoic acid methyl ester, methyl p-aminobenzoate, benzoic acid, 4-amino-, methyl ester, p-methoxycarbonyl aniline, 4-methoxycarbonyl aniline, p-aminobenzoic acid methyl ester, methyl aniline-4-carboxylate, 4-carbomethoxyaniline, methyl4-aminobenzoate, benzoic acid, p-amino-, methyl ester
PubChem CID 12082
IUPAC Name methyl 4-aminobenzoate
SMILES COC(=O)C1=CC=C(C=C1)N

Specifications

Melting Point 110°C to 114°C
Assay Percent Range 98%
Quantity 25 g
Beilstein 775913
Solubility Information Soluble in alcohol and ether. Slightly soluble in water.
Formula Weight 151.17
Percent Purity 98%
Chemical Name or Material Methyl 4-aminobenzoate
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 210-598-7
RTECSNumber DG2900000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.