Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 4-bromobenzoate, 98+%

Catalog No. aaa1551706
Click to view available options
Quantity:
5 g
25 g
100 g

CAS: 619-42-1 | C8H7BrO2 | 215.046 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
Methyl 4-bromobenzoate is used as a starting material in the synthesis of three-carbon-bridged 5-substituted furo[2,3-d]pyrimidine and 6-substituted pyrrolo[2,3-d]pyrimidine and methyl 4-tri-n-butylstannylbenzoate. It is also used to stimulate microbial dechlorination of polychlorinated biphenyls. Further, it is used in the preparation of stable radioiodinating reagent to label monoclonal antibodies for radiotherapy of cancer.

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 619-42-1
Molecular Formula C8H7BrO2
Molecular Weight (g/mol) 215.046
MDL Number MFCD00013531
InChI Key CZNGTXVOZOWWKM-UHFFFAOYSA-N
Synonym benzoic acid, 4-bromo-, methyl ester, methyl p-bromobenzoate, 4-bromobenzoic acid methyl ester, p-bromobenzoic acid, methyl ester, unii-ub1ysj5xar, ub1ysj5xar, benzoic acid, p-bromo-, methyl ester, pubchem3962, methyl-4-bromobenzoate, methyl p-bromo benzoate
PubChem CID 12081
IUPAC Name methyl 4-bromobenzoate
SMILES COC(=O)C1=CC=C(C=C1)Br

Specifications

Melting Point 77°C to 81°C
Density 1.69
Quantity 5 g
Beilstein 2045132
Merck Index 14,1408
Solubility Information Insoluble in water.
Formula Weight 215.05
Percent Purity ≥98%
Chemical Name or Material Methyl 4-bromobenzoate
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 210-596-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.