Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 4-(Bromomethyl)-3-methoxybenzoate 97.0+%, TCI America™
SDP

Catalog No. M235625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M235625G 25 g
M23565G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M235625G Supplier TCI America Supplier No. M235625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 70264-94-7
Molecular Formula C10H11BrO3
Molecular Weight (g/mol) 259.099
MDL Number MFCD00270115
InChI Key UXSNXOMMJXTFEG-UHFFFAOYSA-N
Synonym 4-(Bromomethyl)-3-methoxybenzoic Acid Methyl Ester, Methyl 4-(Bromomethyl)-m-anisate, 4-(Bromomethyl)-m-anisic Acid Methyl Ester
PubChem CID 2759828
IUPAC Name methyl 4-(bromomethyl)-3-methoxybenzoate
SMILES COC1=C(C=CC(=C1)C(=O)OC)CBr

Specifications

Melting Point 93°C
Color White-Yellow
Quantity 25 g
UN Number 3077
Formula Weight 259.10
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 4-(Bromomethyl)-3-methoxybenzoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.