Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 4-(bromomethyl)benzoate, 98%

Catalog No. AAA1055714
Click to view available options
Quantity:
5 g
25 g
100 g

CAS: 2417-72-3 | C9H9BrO2 | 229.07 g/mol

Methyl 4-(bromomethyl)benzoate is used in the preparation of potential anti-HIV agents. It i s also used as a catalyst for rearrangement of benzylthiothiazoline derivatives in the preparation aldose reductase inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 4-(bromomethyl)benzoate is used in the preparation of potential anti-HIV agents. It i s also used as a catalyst for rearrangement of benzylthiothiazoline derivatives in the preparation aldose reductase inhibitors.

Solubility
Lowly soluble in water, highly soluble in ethanol and ether, not soluble in halogenated organic solvents

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Air sensitive. Handle and store under inert gas. Keep away from strong oxidizing agents, strong bases. Stable under recommended storage conditions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2417-72-3
Molecular Formula C9H9BrO2
Molecular Weight (g/mol) 229.07
MDL Number MFCD00032453
InChI Key NLWBJPPMPLPZIE-UHFFFAOYSA-N
PubChem CID 256687
IUPAC Name methyl 4-(bromomethyl)benzoate
SMILES COC(=O)C1=CC=C(CBr)C=C1

Specifications

Melting Point 53°C to 58°C
Boiling Point 130°C to 135°C (2 mmHg)
Quantity 25 g
UN Number UN1759
Beilstein 1867272
Synonym methyl 4-bromomethyl benzoate, 4-bromomethylbenzoic acid methyl ester, methyl4-bromomethyl benzoate, benzoic acid, 4-bromomethyl-, methyl ester, 4-bromomethyl benzoic acid methyl ester, p-methoxycarbonyl benzyl bromide, methyl p-bromomethyl benzoate, 4-bromomethyl-benzoic acid methyl ester, 4-carbomethoxybenzyl bromide
Solubility Information Lowly soluble in water,highly soluble in ethanol and ether,not soluble in halogenated organic solvents
Formula Weight 229.07
Percent Purity 98%
Chemical Name or Material Methyl 4- (bromomethyl)benzoate
Hazard Category H302-H314-H334-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P270-P271-P280-P285-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
EINECSNumber 000-000-0
TSCA No

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.