Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 4-Chloro-2-methylbutyrate 98.0+%, TCI America™
SDP

Catalog No. C131925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C131925G 25 g
C13195G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C131925G Supplier TCI America Supplier No. C131925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 13888-03-4
Molecular Formula C6H11ClO2
Molecular Weight (g/mol) 150.602
MDL Number MFCD00198001
InChI Key ZDDYJHVTUSUPFC-UHFFFAOYSA-N
Synonym 4-Chloro-2-methylbutyric Acid Methyl Ester
PubChem CID 11263584
IUPAC Name methyl 4-chloro-2-methylbutanoate
SMILES CC(CCCl)C(=O)OC

Specifications

Color Colorless
Boiling Point 81°C
Quantity 25 g
Formula Weight 150.60
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl 4-Chloro-2-methylbutyrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.