Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 4-chlorobutyrate, 98+%, ACROS Organics™

Click to view available options
Quantity:
5g
100g
Packaging:
Glass bottle
3153-37-5
C5H9ClO2
136.58
MFCD00001003
ZZUYIRISBMWFMV-UHFFFAOYSA-N
methyl 4-chlorobutyrate,butanoic acid, 4-chloro-, methyl ester,4-chlorobutyric acid methyl ester,butyric acid, 4-chloro-, methyl ester,methyl .omega.-chlorobutyrate,4-chlorobutanoic acid methyl ester,.gamma.-chlorobutyric acid methyl ester,methyl-4-chlorbutanoat,gamma-chlorobutyric acid methyl ester,methyl omega-chlorobutyrate
76612
methyl 4-chlorobutanoate
COC(=O)CCCCl
3153-37-5
C5H9ClO2
136.58
MFCD00001003
ZZUYIRISBMWFMV-UHFFFAOYSA-N
methyl 4-chlorobutyrate,butanoic acid, 4-chloro-, methyl ester,4-chlorobutyric acid methyl ester,butyric acid, 4-chloro-, methyl ester,methyl .omega.-chlorobutyrate,4-chlorobutanoic acid methyl ester,.gamma.-chlorobutyric acid methyl ester,methyl-4-chlorbutanoat,gamma-chlorobutyric acid methyl ester,methyl omega-chlorobutyrate
76612
methyl 4-chlorobutanoate
COC(=O)CCCCl
Specific Gravity 1.12
Infrared Spectrum Authentic
Boiling Point 175.0°C to 176.0°C
Packaging Glass bottle
Density 1.1200g/mL
Quantity 100g
Color Colorless
Refractive Index 1.4311 to 1.4331
Flash Point 59°C
Assay Percent Range 98+%
Linear Formula Cl(CH2)3CO2CH3
Beilstein 02,278
Solubility Information Solubility in water: .. Other solubilities: soluble in organic solvents
Formula Weight 136.58
Physical Form Liquid
Percent Purity ≥98%
Chemical Name or Material Methyl 4-chlorobutyrate, 98+%
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 221-592-9

TSCA : TSCA