Alfa Aesar™ Methyl 4-ethynylbenzoate, 97%

Manufacturer: Alfa Aesar™ H5341906

Quantity 5g

 View more versions of this product

Catalog No. AAH5341906

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 3034-86-4
    Molecular Formula C10H8O2
    Formula Weight 160.17
    MDL Number MFCD00168820
    Synonym 4-Ethynylbenzoic acid methyl ester
    Chemical Name or Material Methyl 4-ethynylbenzoate

    Specifications

    Formula Weight 160.17
    Molecular Formula C10H8O2
    MDL Number MFCD00168820
    Melting Point 92° to 94°C
    Quantity 5g
    CAS 3034-86-4
    Synonym 4-Ethynylbenzoic acid methyl ester
    Chemical Name or Material Methyl 4-ethynylbenzoate
    Assay 97%

    Description

    Applications
    Methyl 4-ethynylbenzoate is used as pharmaceutical intermediate.

    Solubility
    Sparingly soluble in water (0.16 g/L at 25°C).

    Notes
    Stable under recommended storage conditions. Incompatible with oxidizing agents.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit