Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 4-hydroxy-3-iodobenzoate, 98%

Catalog No. aah5687706
Click to view available options
Quantity:
5 g

CAS: 15126-06-4 | C8H7IO3 | 278.045 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15126-06-4
Molecular Formula C8H7IO3
Molecular Weight (g/mol) 278.045
MDL Number MFCD05664378
InChI Key PXNOLLHARLSLHY-UHFFFAOYSA-N
Synonym methyl4-hydroxy-3-iodobenzoate, methyl 3-iodo-4-hydroxybenzoate, methyl 4-hydroxy-3-iodo-benzoate, 4-hydroxy-3-iodobenzoic acid methyl ester, 4-hydroxy-3-iodo-benzoic acid methyl ester, benzoic acid, 4-hydroxy-3-iodo-, methyl ester, 4-hydroxy-3-iodo-benzoicacidmethylester, acmc-209zk0, ksc495e2f, methyl 4-hydroxy-3-iodbenzoate
PubChem CID 11346465
IUPAC Name methyl 4-hydroxy-3-iodobenzoate
SMILES COC(=O)C1=CC(=C(C=C1)O)I

Specifications

Melting Point 155°C to 159°C
Color Brown to White
Quantity 5 g
Sensitivity Light sensitive
Formula Weight 278.04
Percent Purity 98%
Physical Form Crystals or Powder
Chemical Name or Material Methyl 4-hydroxy-3-iodobenzoate
Hazard Category H302-H318
Hazard Statement GHS H Statement
H318-H302
Causes serious eye damage.
Harmful if swallowed.
Precautionary Statement P264b-P270-P280-P301+P312-P305+P351+P338-P310-P330-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.