Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 4-hydroxy-3-nitrobenzoate, 98+%

Catalog No. p-7081064
Click to view available options
Quantity:
50 g
250 g

CAS: 99-42-3 | C8H7NO5 | 197.146 g/mol

4-alkoxy-3-nitrobenzoic acids were obtained by the alkylation of methyl 4-hydroxy-3-nitrobenzoate with corresponding n-alkylbromides in 3-pentanone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-alkoxy-3-nitrobenzoic acids were obtained by the alkylation of methyl 4-hydroxy-3-nitrobenzoate with corresponding n-alkylbromides in 3-pentanone.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99-42-3
Molecular Formula C8H7NO5
Molecular Weight (g/mol) 197.146
MDL Number MFCD00014695
InChI Key GNCWCTBHZCBXGL-UHFFFAOYSA-N
Synonym methyl 3-nitro-4-hydroxybenzoate, benzoic acid, 4-hydroxy-3-nitro-, methyl ester, 4-hydroxy-3-nitrobenzoic acid methyl ester, methyl 3-nitro-4-hydroxy toluate, aurora 6897, 3-nitro-4-hydroxybenzoic acid methyl ester, methyl-4-hydroxy-3-nitrobenzoic acid, 3-nitro-4-hydroxymethylbenzoate, acmc-209scn, 3-10-00-00378 beilstein handbook reference
PubChem CID 66835
IUPAC Name methyl 4-hydroxy-3-nitrobenzoate
SMILES COC(=O)C1=CC(=C(C=C1)O)[N+](=O)[O-]

Specifications

Melting Point 73°C to 77°C
Quantity 50 g
Beilstein 2115197
Formula Weight 197.15
Percent Purity ≥98%
Chemical Name or Material Methyl 4-hydroxy-3-nitrobenzoate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 202-755-3
RTECSNumber DH2528400
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.