Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 4-hydroxybenzoate, sodium salt, 99%, Thermo Scientific Chemicals

Catalog No. p-164300
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Plastic bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5026-62-0
Molecular Formula C8H7NaO3
Molecular Weight (g/mol) 174.13
MDL Number MFCD00016470
InChI Key PESXGULMKCKJCC-UHFFFAOYSA-M
Synonym methylparaben sodium, sodium methylparaben, sodium 4-methoxycarbonyl phenolate, preserval ms, methyl 4-hydroxybenzoate, sodium salt, solparol, bonomold omna, methyl 4-hydroxybenzoate sodium salt, sodium 4-carbomethoxyphenolate, unii-cr6k9c2nhk
PubChem CID 23663626
IUPAC Name sodium;4-methoxycarbonylphenolate
SMILES COC(=O)C1=CC=C(C=C1)[O-].[Na+]

Specifications

Melting Point >125°C
Color White
Flash Point >100°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Plastic bottle
Quantity 500 g
Merck Index 10,5977
Solubility Information Solubility in water: 333g/L (20°C)
Formula Weight 174.13
Percent Purity 99%
Physical Form Powder
Chemical Name or Material Methyl 4-hydroxybenzoate, sodium salt, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye damage.
Harmful to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
EINECSNumber 225-714-1
RTECSNumber DH2455000

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.