Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Methyl 4-hydroxyindole-6-carboxylate, 97%, Thermo Scientific™
Click to view available options
:
250mg
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
Methyl 4-hydroxyindole-6-carboxylate is used as a pharmaceutical intermediate.
Solubility
Slightly soluble in water (2.5 g/L) (25°C).
Notes
Heat sensitive. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
Methyl 4-hydroxyindole-6-carboxylate is used as a pharmaceutical intermediate.
Solubility
Slightly soluble in water (2.5 g/L) (25°C).
Notes
Heat sensitive. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.

Specifications
Specifications
CAS | 77140-48-8 |
Molecular Formula | C10H9NO3 |
Quantity | 250mg |
MDL Number | MFCD04973985 |
Synonym | 4-hydroxy-1h-indole-6-carboxylic acid methyl ester, 4-hydroxy-6-methoxycarbonyl indole, methyl 4-hydroxyindole-6-carboxylate, 1h-indole-6-carboxylic acid, 4-hydroxy-, methyl ester |
Solubility Information | Slightly soluble in water (2.5g/L) (25°C). |
InChI Key | YCLHTEHHXRJHKC-UHFFFAOYSA-N |
SMILES | COC(=O)C1=CC2=C(C=CN2)C(=C1)O |
IUPAC Name | methyl 4-hydroxy-1H-indole-6-carboxylate |
Molecular Weight (g/mol) | 191.186 |
Show More |
Safety and Handling
TSCA | No |
storageNote1 | 2° to 8°C |
Recommended Storage | Keep cold |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction