Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 4-(hydroxymethyl)pyridine-2-carboxylate, 95%

Catalog No. p-7031600
Click to view available options
Quantity:
1 g
5 g

Catalog No. AAH3273103

Supplier: Thermo Scientific Chemicals H3273103

Only null left
Add to Cart

CAS: 317335-15-2 | C8H9NO3 | 167.164 g/mol

Methyl 4-(hydroxymethyl)pyridine-2 is used as an intermediate in pharmaceuticals and also finds use in chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 4-(hydroxymethyl)pyridine-2 is used as an intermediate in pharmaceuticals and also finds use in chemical research.

Notes
Store in a cool place. Incompatible with acids, bases and strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 317335-15-2
Molecular Formula C8H9NO3
Molecular Weight (g/mol) 167.164
MDL Number MFCD09834350
InChI Key WEIFQBALQLWZFZ-UHFFFAOYSA-N
Synonym methyl 4-hydroxymethyl picolinate, methyl 4-hydroxymethyl pyridine-2-carboxylate, methyl 4-hydroxymethyl-pyridine-2-carboxylate, 4-hydroxymethyl-2-pyridinecarboxylic acid methyl ester, 2-pyridinecarboxylic acid, 4-hydroxymethyl-, methyl ester, 2-pyridinecarboxylicacid,4-hydroxymethyl-,methylester 9ci, 2-methoxycarbonylpyridine-4-methanol, 4-hydroxymethyl-pyridine-2-carboxylic acid methyl, 4-hydroxymethylpyridin-2-carboxylic acid methyl ester, 4-hydroxymethyl pyridine-2-carboxylic acid methyl ester
PubChem CID 23093785
IUPAC Name methyl 4-(hydroxymethyl)pyridine-2-carboxylate
SMILES COC(=O)C1=NC=CC(=C1)CO

Specifications

Melting Point 74°C to 76°C
Quantity 1 g
Formula Weight 167.16
Percent Purity 95%
Chemical Name or Material Methyl 4-(hydroxymethyl)pyridine-2-carboxylate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.