Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 4-hydroxyphenylacetate, 99%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
Packaging:
Glass bottle
14199-15-6
C9H10O3
166.18
MFCD00002387
XGDZEDRBLVIUMX-UHFFFAOYSA-N
methyl 4-hydroxyphenylacetate,methyl 2-4-hydroxyphenyl acetate,4-hydroxyphenylacetic acid methyl ester,benzeneacetic acid, 4-hydroxy-, methyl ester,methyl 4-hydroxyphenyl acetate,4-hydroxyphenylacetic acid methl ester,acetic acid, p-hydroxyphenyl-, methyl ester,p-hydroxyphenylacetic acid methyl ester,4-hydroxybenzeneacetic acid methyl ester,4-hydroxy-phenyl-acetic acid methyl ester
518900
CHEBI:68078
methyl 2-(4-hydroxyphenyl)acetate
COC(=O)CC1=CC=C(C=C1)O
14199-15-6
C9H10O3
166.18
MFCD00002387
XGDZEDRBLVIUMX-UHFFFAOYSA-N
methyl 4-hydroxyphenylacetate,methyl 2-4-hydroxyphenyl acetate,4-hydroxyphenylacetic acid methyl ester,benzeneacetic acid, 4-hydroxy-, methyl ester,methyl 4-hydroxyphenyl acetate,4-hydroxyphenylacetic acid methl ester,acetic acid, p-hydroxyphenyl-, methyl ester,p-hydroxyphenylacetic acid methyl ester,4-hydroxybenzeneacetic acid methyl ester,4-hydroxy-phenyl-acetic acid methyl ester
518900
CHEBI:68078
methyl 2-(4-hydroxyphenyl)acetate
COC(=O)CC1=CC=C(C=C1)O
Quantity 100g
Melting Point 55°C to 58°C
Infrared Spectrum Authentic
Packaging Glass bottle
Boiling Point 162°C to 163°C (5.0mmHg)
Color White to Yellow
Assay Percent Range 99%
Linear Formula HOC6H4CH2CO2CH3
Beilstein 10, 191
Formula Weight 166.18
Physical Form Crystals
Percent Purity 99%
Chemical Name or Material Methyl 4-hydroxyphenylacetate, 99%
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 238-050-2

TSCA : TSCA