Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 5-chloro-2-methoxybenzoate, 98%

Catalog No. AAA1078722
Click to view available options
Quantity:
25 g
100 g
500 g

CAS: 33924-48-0 | C9H9ClO3 | 200.618 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 33924-48-0
Molecular Formula C9H9ClO3
Molecular Weight (g/mol) 200.618
MDL Number MFCD00017511
InChI Key HPTHYBXMNNGQEF-UHFFFAOYSA-N
Synonym methyl 5-chloro-o-anisate, methyl-5-chloro-2-methoxybenzoate, 5-chloro-2-methoxybenzoic acid methyl ester, benzoic acid, 5-chloro-2-methoxy-, methyl ester, 5-chloro-o-anisic acid methyl ester, 5-chloro-2-methoxy-benzoic acid methyl ester, pubchem3709, methyl5-chloro-2-methoxybenzoate, acmc-1agpb, dsstox_cid_31509
PubChem CID 118574
IUPAC Name methyl 5-chloro-2-methoxybenzoate
SMILES COC1=C(C=C(C=C1)Cl)C(=O)OC

Specifications

Density 1.259
Boiling Point 235°C to 240°C
Flash Point >110°C (230°F)
Refractive Index 1.546
Quantity 100 g
Beilstein 2719088
Formula Weight 200.62
Percent Purity 98%
Chemical Name or Material Methyl 5-chloro-2-methoxybenzoate
EINECSNumber 251-743-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.