Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
α-Methyl-5-hydroxytryptamine maleate, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
5-HT2B receptor selective agonist with pKi values of 8.4, 6.1 and 7.3 at 5-HT2B, 5-HT2A and 5-HT2C receptors respectively.
Specifications
Specifications
Quantity | 50 mg |
Formula Weight | 306.32 |
Percent Purity | >99% |
Chemical Name or Material | α-Methyl-5-hydroxytryptamine maleate |
Safety and Handling
Recommended Storage | Store at -20°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction