Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 5-iodosalicylate, 99%

Catalog No. p-7026189
Click to view available options
Quantity:
10 g
50 g

Catalog No. AAB2204809

Supplier: Thermo Scientific Chemicals B2204809

Only null left
Add to Cart

CAS: 4068-75-1 | C8H7IO3 | 278.045 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4068-75-1
Molecular Formula C8H7IO3
Molecular Weight (g/mol) 278.045
MDL Number MFCD00016462
InChI Key NRSWJTRJHPRZMH-UHFFFAOYSA-N
PubChem CID 248910
IUPAC Name methyl 2-hydroxy-5-iodobenzoate
SMILES COC(=O)C1=C(C=CC(=C1)I)O

Specifications

Melting Point 75°C to 78°C
Quantity 10 g
Sensitivity Light sensitive
Synonym methyl 5-iodosalicylate, 5-iodosalicylic acid methyl ester, 2-hydroxy-5-iodobenzoic acid methyl ester, benzoic acid, 2-hydroxy-5-iodo-, methyl ester, methyl 2-hydroxy-5-iodobenzenecarboxylate, salicylic acid, 5-iodo-, methyl ester, methyl5-iodosalicylate, methyl-5-iodosalicylate, acmc-209jg0, methyl2-hydroxy-5-iodobenzoate
Formula Weight 278.05
Percent Purity 99%
Chemical Name or Material Methyl 5-iodosalicylate
Hazard Category H301-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.