Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 6-chloropyridine-2-carboxylate, 95%

Catalog No. aah6402306
Click to view available options
Quantity:
5 g
25 g
100 g

CAS: 6636-55-1 | C7H6ClNO2 | 171.58 g/mol

Methyl 6-chloropyridine-2-carboxylate was coupled with 2-(trimethylstannyl)pyridine to afford methyl 2,2?-bipyridine-6-carboxylate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 6-chloropyridine-2-carboxylate was coupled with 2-(trimethylstannyl)pyridine to afford methyl 2,2′-bipyridine-6-carboxylate.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, heat.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6636-55-1
Molecular Formula C7H6ClNO2
Molecular Weight (g/mol) 171.58
MDL Number MFCD00155391
InChI Key TWUXBVMXSBEKHA-UHFFFAOYSA-N
Synonym methyl 6-chloropicolinate, methyl 6-chloro-2-pyridinecarboxylate, 2-pyridinecarboxylic acid, 6-chloro-, methyl ester, 2-chloropyridine-6-carboxylate methyl, 6-chloro-2-picolinic acid methyl ester, 6-chloropicolinic acid methyl ester, methyl 2-chloro-6-pyridinecarboxylate, methyl 2-chloropyridine-6-carboxylate, 6-chloro-pyridine-2-carboxylic acid methyl ester, 6-chloro-2-pyridinecarboxylic acid methyl ester
PubChem CID 242723
IUPAC Name methyl 6-chloropyridine-2-carboxylate
SMILES COC(=O)C1=NC(=CC=C1)Cl

Specifications

Melting Point 92°C to 98°C (literature)
Color Yellow
Quantity 5 g
Beilstein 128580
Solubility Information Slightly soluble in water.
Formula Weight 171.58
Percent Purity 95%
Physical Form Solid
Chemical Name or Material Methyl 6-chloropyridine-2-carboxylate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.