Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

methyl 6-phenylnicotinate, Thermo Scientific™

Catalog No. CC21122DA
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC21122DA 1 g
CC21122DE 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC21122DA Supplier Thermo Scientific Chemicals Supplier No. CC21122DA

Chemical Identifiers

CAS 4634-13-3
Molecular Formula C13H11NO2
Molecular Weight (g/mol) 213.236
InChI Key DXHZSBAEWGPUKI-UHFFFAOYSA-N
Synonym methyl 6-phenylnicotinate, methyl 6-phenyl-3-pyridinecarboxylate, 6-phenylpyridine-3-carboxylic acid methyl ester
PubChem CID 13129505
IUPAC Name methyl 6-phenylpyridine-3-carboxylate
SMILES COC(=O)C1=CN=C(C=C1)C2=CC=CC=C2

Specifications

Assay Percent Range 97%
Quantity 1 g
Formula Weight 213.23
Percent Purity 97%
Chemical Name or Material Methyl 6-phenylnicotinate
Hazard Category GHS Signal Word: Warning
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.