Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl Acetate (Certified ACS), Fisher Chemical™

Catalog No. p-158815 Shop All Fisherbrand Products
Click to view available options
Quantity:
500 mL
4 L
Packaging:
Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 79-20-9
Molecular Formula C3H6O2
Molecular Weight (g/mol) 74.079
MDL Number MFCD00008711
InChI Key KXKVLQRXCPHEJC-UHFFFAOYSA-N
Synonym tereton, devoton, acetic acid, methyl ester, methyl ethanoate, acetic acid methyl ester, methylacetat, acetate de methyle, methylacetaat, methyl acetic ester, octan metylu
PubChem CID 6584
ChEBI CHEBI:77700
IUPAC Name methyl acetate
SMILES CC(=O)OC

Specifications

Boiling Range 1.0°C max.
Melting Point -98°C
Color Colorless
Density 0.930g/cm³
Boiling Point 57.4°C
Flash Point −26°C
Packaging Glass Bottle
Linear Formula CH3COOCH3
Quantity 500 mL
UN Number 1231
Residue after Evaporation 0.002% max.
Specific Gravity 0.933 to 0.938 (at 20°C)
Formula Weight 74.08
Grade Certified
Physical Form Liquid
Chemical Name or Material Methyl Acetate
Show More Show Less
DOTInformation DOT Class 3, : Flammable Liquid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.