Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl Alcohol, Exceeds A.C.S Specifications, HPLC Grade, 99.9%, Spectrum™ Chemical
SDP

Catalog No. 18604409 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
1 L
4 L
4 x 4 L
6 x 1 L
Packaging:
Amber Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-604-409 1 L Amber Glass Bottle
18-604-410 4 L Amber Glass Bottle
18-604-411 6 x 1 L Amber Glass Bottle
18-604-412 4 x 4 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-604-409 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. HP7021LT
Add to Cart
Add to Cart

HP702, 67-56-1, CH3OH

Spectrum™ Chemical Methyl Alcohol, Exceeds A.C.S Specifications, HPLC Grade is a colorless, flammable substance used in chemical labs as a polar protic solvent in organic synthesis, as well as a carrier (mobile phase) solvent in high pressure liquid chromatography (HPLC). Less expensive than Acetonitrile, the popularity Methyl Alcohol HPLC continues to grow, especially in reversed phase HPLC.

All HPLC and Ultra-pure solvents offered by Spectrum are glass distilled, submicron filtered, and undergo rigorous specification testing to insure accurate, repeatable results.

Chemical Identifiers

CAS 67-56-1
Molecular Formula CH4O
Molecular Weight (g/mol) 32.04
MDL Number MFCD00004595
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
IUPAC Name methanol
SMILES CO

Specifications

CAS Max % 100%
Quantity 1 L
Packaging Amber Glass Bottle
Residue after Evaporation 2 ppm
Titratable Acid 0.0003 meq/g
Titratable Base 0.0002 meq/g
Water 0.05%
UN Number UN1230
Boiling Point 64°C to 65°C
Percent Purity 99.9%
Grade High Purity
Chemical Name or Material Methyl Alcohol
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.