Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl Benzoylformate 97.0+%, TCI America™
SDP

Catalog No. B1033100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B1033100G 100 g
B103325G 25 g
B1033500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B1033100G Supplier TCI America Supplier No. B1033100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 15206-55-0
Molecular Formula C9H8O3
Molecular Weight (g/mol) 164.16
MDL Number MFCD00008443
InChI Key YLHXLHGIAMFFBU-UHFFFAOYSA-N
Synonym methyl benzoylformate, methyl phenylglyoxylate, methyl oxophenylacetate, methyl phenylglyoxalate, benzoylformic acid methyl ester, methylbenzoylformate, phenylglyoxylic acid methyl ester, phenylglyoxylic acid, methyl ester, benzeneacetic acid, .alpha.-oxo-, methyl ester, unii-23f2045stg
PubChem CID 84835
ChEBI CHEBI:84256
IUPAC Name methyl 2-oxo-2-phenylacetate
SMILES COC(=O)C(=O)C1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 251°C
Quantity 100 g
Formula Weight 164.16
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl Benzoylformate
EINECSNumber (3)-3690
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.