Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl-beta-cyclodextrin

Catalog No. p-7057555
Click to view available options
Quantity:
5 g
25 g

CAS: 128446-36-6 | C54H94O35 | 1303.311 g/mol

Cavity size is the major determinant as to which cyclodextrin is used in complexation. The cavity diameter of β-cyclodextrins or β-glucopyranose unit compounds is well-suited for use with molecules the size of hormones, vitamins and many compounds frequently used in tissue and cell culture applications. For this reason, β-cyclodextrin is most commonly used as a complexing agent. It is used to increase the solubility of non-polar substances such as fatty acids, lipids, vitamins and cholesterol for use in cell culture applications. Methyl-β-cyclodextrin is used to remove cholesterol from cultured cells and is employed to remove lipid rafts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cavity size is the major determinant as to which cyclodextrin is used in complexation. The cavity diameter of β-cyclodextrins or β-glucopyranose unit compounds is well-suited for use with molecules the size of hormones, vitamins and many compounds frequently used in tissue and cell culture applications. For this reason, β-cyclodextrin is most commonly used as a complexing agent. It is used to increase the solubility of non-polar substances such as fatty acids, lipids, vitamins and cholesterol for use in cell culture applications. Methyl-β-cyclodextrin is used to remove cholesterol from cultured cells and is employed to remove lipid rafts.

Solubility
Soluble in water

Notes
Store at room temperature. Incompatible with heat, oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 128446-36-6
Molecular Formula C54H94O35
Molecular Weight (g/mol) 1303.311
MDL Number MFCD00074980
InChI Key YZOUYRAONFXZSI-SBHWVFSVSA-N
Synonym beta-cyclodextrin methyl ethers, beta-cyd, methyl-?cyclodextrin, beta-w7 m1.8, dimethyl-, a-cyclodextrin, .beta.-cyclodextrin,methylethers, methyl-b-cyclodextrin cell culture*tested
PubChem CID 51051622
SMILES COC1C(C2C(OC1OC3C(OC(C(C3OC)OC)OC4C(OC(C(C4OC)OC)OC5C(OC(C(C5OC)OC)OC6C(OC(C(C6OC)OC)OC7C(OC(C(C7OC)OC)OC8C(OC(O2)C(C8O)OC)CO)CO)CO)CO)CO)CO)CO)O

Specifications

Melting Point 180°C to 182°C
Color White
Quantity 5 g
Solubility Information Soluble in water
Physical Form Powder
Chemical Name or Material Methyl-beta-cyclodextrin
Hazard Category H317-H361
Precautionary Statement P201-P202-P261-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P501c
EINECSNumber 411-120-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.