Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl blue, MP Biomedicals™

Catalog No. ICN15264325 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15264325 25 g
ICN15264305 5 g
ICN215401 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN15264325 Supplier MP Biomedicals, Inc Supplier No. 0215264325
Add to Cart
Add to Cart

Chemical Identifiers

CAS 28631-66-5
Molecular Formula C32H27N3
Molecular Weight (g/mol) 453.59
MDL Number MFCD00036130
InChI Key LBLFVSANTXZPEX-UHFFFAOYSA-N
Synonym Acid blue 93, Aniline blue
PubChem CID 131675892
IUPAC Name 2-methyl-4-{[4-(phenylamino)phenyl][4-(phenylimino)cyclohexa-2,5-dien-1-ylidene]methyl}aniline
SMILES CC1=CC(=CC=C1N)C(C1=CC=C(NC2=CC=CC=C2)C=C1)=C1C=CC(C=C1)=NC1=CC=CC=C1

Specifications

Formula Weight 799.8
Color Black/Purple to Blue
Quantity 25 g
Physical Form Powder
Chemical Name or Material Methyl blue
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.