Learn More
Iodoacetic acid, sodium salt, 99%
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 208.0°C to 210.0°C |
| Color | Brown-Yellow to Yellow |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 99% |
| Packaging | Glass bottle |
| Linear Formula | ICH2CO2Na |
| Quantity | 100 g |
| Beilstein | 02, 222 |
| Fieser | 13,116 |
| Solubility Information | Solubility in water: soluble. Other solubilities: soluble in alcohol,slightly soluble in ether |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement Toxic if swallowed. May cause respiratory irritation. Causes skin irritation. Causes serious eye damage. |
| Precautionary Statement | GHS P Statement IF ON SKIN: Wash with plenty of soap and water. If skin irritation occurs: Get medical advice/attention. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses,if present and easy to do. |
| EINECSNumber | 206-165-7 |
| RTECSNumber | AI3675000 |
| TSCA | TSCA |
| Recommended Storage | −20°C |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.