Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silver sulfate, pure, 10 g/l solution in sulfuric acid

Catalog No. AC279480025
Click to view available options
Quantity:
1 L
2.5 L

CAS: 10294-26-5 | Ag2H2O4S | 313.81 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Silver sulfate
Color Colorless
Name Note 10g/L Solution in Sulfuric Acid
CAS 10294-26-5, 7664-93-9
Molecular Formula Ag2H2O4S
Linear Formula Ag2SO4
MDL Number MFCD00003407
Fieser 01,1015; 03,254; 04,435
Merck Index 15, 8668
Density 1.8400g/mL
Synonym silver sulfate, disilver sulfate, disilver 1+ sulfate, unii-8qg6hv4zpo, disilver 1+ sulphate, sulfuric acid, disilver 1+ salt, 8qg6hv4zpo, disilver 1+ ion sulfate, sulfuric acid, silver 1+ salt 1:2, sulfuric acid disilver i salt
InChI Key YPNVIBVEFVRZPJ-UHFFFAOYSA-N
SMILES [Ag+].[Ag+].OS(O)(=O)=O
IUPAC Name disilver(1+) sulfuric acid
Molecular Weight (g/mol) 313.81
PubChem CID 159865
Formula Weight 311.79
Grade Pure
Packaging Glass bottle
Specific Gravity 1.84
Quantity 2.5 L
Physical Form Liquid
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.