Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl cholate, 97%

Catalog No. p-7044192
Click to view available options
Quantity:
5 g
25 g

CAS: 1448-36-8 | C25H42O5 | 422.606 g/mol

Methyl cholate, is used as a bile acid derivative as anti-inflammatory and antitumor agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl cholate, is used as a bile acid derivative as anti-inflammatory and antitumor agent.

Solubility
Insoluble in water. Soluble in medium polarity organic solvents.

Notes
Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1448-36-8
Molecular Formula C25H42O5
Molecular Weight (g/mol) 422.606
MDL Number MFCD00064934
InChI Key DLYVTEULDNMQAR-BSEHQKOQSA-N
Synonym Cholic acid methyl ester
PubChem CID 133124937
IUPAC Name methyl (4S)-4-[(3R,7S,8S,9R,10R,12R,13S,14R,17S)-3,7,12-trihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoate
SMILES CC(CCC(=O)OC)C1CCC2C1(C(CC3C2C(CC4C3(CCC(C4)O)C)O)O)C

Specifications

Melting Point 155°C to 156°C
Quantity 5 g
Beilstein 2226989
Merck Index 14,2203
Solubility Information Insoluble in water. Soluble in medium polarity organic solvents.
Formula Weight 422.61
Percent Purity ≥98%
Chemical Name or Material Methyl cholate
EINECSNumber 215-903-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.