Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl cinnamate, predominantly trans, 99%

Catalog No. aaa1597522
Click to view available options
Quantity:
100 g
500 g
2500 g

CAS: 103-26-4 | C10H10O2 | 162.188 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
Methyl cinnamate was used to inhibit monophenolase and diphenolase activity of mushroom tyrosinase and it also has antimicrobial ability. It is mainly is used in the flavor and perfume industries. It is known to attract males of various orchid bees, such as Aglae caerulea.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103-26-4
Molecular Formula C10H10O2
Molecular Weight (g/mol) 162.188
MDL Number MFCD00008458
InChI Key CCRCUPLGCSFEDV-BQYQJAHWSA-N
Synonym methyl cinnamate, methyl trans-cinnamate, methyl e-cinnamate, cinnamic acid methyl ester, methyl cinnamylate, methyl 3-phenylacrylate, methyl e-3-phenylprop-2-enoate, methyl 3-phenylpropenoate, trans-cinnamic acid methyl ester, trans-methyl cinnamate
PubChem CID 637520
ChEBI CHEBI:6857
IUPAC Name methyl (E)-3-phenylprop-2-enoate
SMILES COC(=O)C=CC1=CC=CC=C1

Specifications

Melting Point 34°C to 37°C
Density 1.042
Boiling Point 262°C to 264°C
Flash Point 123°C (253°F)
Assay Percent Range 99%
Refractive Index 1.577
Quantity 100 g
Beilstein 386468
Merck Index 14,2299
Solubility Information Insoluble in water.
Formula Weight 162.19
Percent Purity 99%
Chemical Name or Material Methyl Cinnamate, predominantly trans
Show More Show Less
EINECSNumber 203-093-8
RTECSNumber GE0190000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.