Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl cis-13-docosenoate, ≥99.0% (GC), MilliporeSigma™ Supelco™

Catalog No. p-7187778
Click to view available options
Quantity:
1 mL
5 mL

Specifications

CAS 122-34-9
Density 0.870 g/mL (at 20°C)
Flash Point 33°C
Shipping Condition Ambient
Physical Form Neat
Linear Formula CH3(CH2)7 CH=CH(CH2)11 COOCH3
MDL Number MFCD00027343
Boiling Point 172°C to 175°C (0.2 mmHg)
Synonym Methyl erucate
Type Analytical Standard
Quantity 1 mL
Formula Weight 352.59
Percent Purity ≥99.0% (GC)
Grade Analytical Standard
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.