Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl dichloromethoxyacetate, 90%, Tech., Thermo Scientific™

Catalog No. AC243560250
Click to view available options
Quantity:
25 mL
Packaging:
Glass bottle

Chemical Identifiers

CAS 17640-25-4
Molecular Formula C4H6Cl2O3
Molecular Weight (g/mol) 173
MDL Number MFCD00012202
InChI Key FWIGIASHGJFYOL-UHFFFAOYSA-N
Synonym methyldichloromethoxyacetate, methyl2,2-dichloro-2-methoxyacetate, methoxydichloroacetic acid methyl ester, methyl 2,2-dichloro-2-methoxyacetate, methyl 2 pound not2-dichloro-2-methoxyacetate, aceticacid,2,2-dichloro-2-methoxy-,methylester, acetic acid,2,2-dichloro-2-methoxy-,methyl ester
PubChem CID 2724502
IUPAC Name methyl 2,2-dichloro-2-methoxyacetate
SMILES COC(=O)C(OC)(Cl)Cl

Specifications

Color Colorless
Density 1.3400g/mL
Boiling Point 181.0°C
Flash Point 91°C
Infrared Spectrum Authentic
Assay Percent Range 88% min. (GC)
Packaging Glass bottle
Refractive Index 1.4496 to 1.4516
Quantity 25 mL
Specific Gravity 1.34
Formula Weight 173
Percent Purity 90%
Grade Technical
Physical Form Liquid
Chemical Name or Material Methyl dichloromethoxyacetate, tech., 90%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF ON SKIN (

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.