Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl Dimethoxyacetate 95.0+%, TCI America™
SDP

Catalog No. D050310ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D050310ML 10 mL
D050325ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D050310ML Supplier TCI America Supplier No. D050310ML

Chemical Identifiers

CAS 89-91-8
Molecular Formula C5H10O4
Molecular Weight (g/mol) 134.131
MDL Number MFCD00008484
InChI Key NZTCVGHPDWAALP-UHFFFAOYSA-N
Synonym methyl dimethoxyacetate, acetic acid, dimethoxy-, methyl ester, dimethoxyacetic acid methyl ester, glyoxylic acid methyl ester dimethyl acetal, acetic acid, 2,2-dimethoxy-, methyl ester, glyoxylic acid, methyl ester, 2-dimethyl acetal, zlchem 519, acmc-209r3k, ksc490c4f, methyl 2,2-dimethoxyethanoate
PubChem CID 66647
IUPAC Name methyl 2,2-dimethoxyacetate
SMILES COC(C(=O)OC)OC

Specifications

Color Yellow
Boiling Point 67°C
Quantity 10 mL
Formula Weight 134.13
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl Dimethoxyacetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.