Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl disulfide, 99%

Catalog No. p-3736920
Click to view available options
Quantity:
50 g
250 g
1 kg
2.5 kg
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

CAS: 624-92-0 | C2H6S2 | 94.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 624-92-0
Molecular Formula C2H6S2
Molecular Weight (g/mol) 94.19
MDL Number MFCD00008561
InChI Key WQOXQRCZOLPYPM-UHFFFAOYSA-N
Synonym dimethyl disulfide, methyl disulfide, dimethyldisulfide, disulfide, dimethyl, dmds, 2,3-dithiabutane, dimethyl disulphide, methyldisulfanyl methane, methyldisulfide, methyldithiomethane
PubChem CID 12232
ChEBI CHEBI:4608
IUPAC Name (methyldisulfanyl)methane
SMILES CSSC

Specifications

Melting Point -85.0°C
Color Yellow
Density 1.0625g/mL
Boiling Point 108.0°C to 110.0°C
Flash Point 16°C
Infrared Spectrum Authentic
Assay Percent Range 1% max. (Methylmercaptan content) (GC)
Packaging Glass bottle
Refractive Index 1.5243 to 1.5263
Linear Formula (CH3)2S2
Quantity 50 g
Beilstein 01, 291
Fieser 05,246; 06,217; 08,189
Specific Gravity 1.0625
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ether,ethyl alcohol,ccl4,benzene
Viscosity 0.620 mPa.s (20°C)
Formula Weight 94.19
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Methyl disulfide
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Highly flammable liquid and vapour.
Toxic if swallowed.
Toxic if inhaled.
May cause an allergic skin reaction.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
I
EINECSNumber 210-871-

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.