Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl dodecanoate, 99%

Catalog No. p-7020509
Click to view available options
Quantity:
5 g
25 g
100 g

CAS: 111-82-0 | C13H26O2 | 214.349 g/mol

Methyl dodecanoate used in production of biodiesel. It has been used in selective synthesis of the secondary amide surfactant, N-methyl lauroylethanolamide. It is also utilized as emollients, perfuming agents skin conditioning.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl dodecanoate used in production of biodiesel. It has been used in selective synthesis of the secondary amide surfactant, N-methyl lauroylethanolamide. It is also utilized as emollients, perfuming agents skin conditioning.

Solubility
Not miscible in water. Soluble in alcohol, dipropylene glycol.

Notes
Store at room temperature. Ensure good ventilation.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-82-0
Molecular Formula C13H26O2
Molecular Weight (g/mol) 214.349
MDL Number MFCD00008966
InChI Key UQDUPQYQJKYHQI-UHFFFAOYSA-N
Synonym methyl laurate, lauric acid methyl ester, dodecanoic acid, methyl ester, methyl laurinate, methyl dodecylate, methyl n-dodecanoate, uniphat a40, metholene 2296, lauric acid, methyl ester, stepan c40
PubChem CID 8139
ChEBI CHEBI:87494
IUPAC Name methyl dodecanoate
SMILES CCCCCCCCCCCC(=O)OC

Specifications

Melting Point 4°C to 5°C
Density 0.870
Boiling Point 261°C to 262°C
Flash Point 125°C (257°F)
Assay Percent Range 99%
Refractive Index 1.4325
Quantity 5 g
Beilstein 1767780
Solubility Information Not miscible in water. Soluble in alcohol,dipropylene glycol.
Formula Weight 214.35
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Methyl Laurate
Show More Show Less
DOTInformation Hazard Class: 9; Packaging Group: III
EINECSNumber 203-911-3
RTECSNumber OF0670000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.