Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl Ethyl Ketone, B&J Brand™, for HPLC, GC and spectrophotometry, >99.5%, Honeywell Burdick & Jackson

Catalog No. 6000119 Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
4 x 4 L
Packaging:
Glass Bottle

Chemical Identifiers

CAS 78-93-3
Molecular Formula C4H8O
Molecular Weight (g/mol) 72.11
MDL Number MFCD00011648
InChI Key ZWEHNKRNPOVVGH-UHFFFAOYSA-N
Synonym 2-butanone, methyl ethyl ketone, butanone, ethyl methyl ketone, methylethyl ketone, meetco, methyl acetone, methylethylketone, 3-butanone, ethylmethylketon
PubChem CID 6569
ChEBI CHEBI:28398
IUPAC Name butan-2-one
SMILES CCC(C)=O

Specifications

Absorbance 0.1 AU max. at 340 nm, 0.02 AU max. at 350 nm, 0.01 AU max. at 375 nm, 0.005 AU max. at 400 nm
CAS Max % 100.0000%
Density 0.805 g/cm3
Boiling Point 79.64°C
Quantity 4 x 4 L
Flash Point −9°C
Packaging Glass Bottle
UN Number UN1193
Beilstein 741880
Formula Weight 72.11
Percent Purity >99.5%
Grade HPLC
Chemical Name or Material Methyl Ethyl Ketone, B&J Brand™, for HPLC, GC and spectrophotometry, >99.5%
Show More Show Less
ShelfLife 547 days from date of manufacture
DOTInformation Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Ethyl methyl ketone
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.