Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl Ethyl Ketone/Hexane, 25/75, v/v, Reagents

Catalog No. 01334712
Click to view available options
:
Quantity: 20 L

Methyl Ethyl Ketone/Hexane, 25/75, v/v, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 110-54-3, 78-93-3
Molecular Formula C6H14
Molecular Weight (g/mol) 86.18
MDL Number MFCD02179311
InChI Key VLKZOEOYAKHREP-UHFFFAOYSA-N
Synonym Methyl Ethyl Ketone/Hexanes
IUPAC Name hexane
SMILES CCCCCC

Specifications

Color Colorless
Quantity 20 L
Packaging Steel Pail
Physical Form Liquid
Specific Gravity 0.6963
Grade Reagent
Chemical Name or Material Methyl Ethyl Ketone/Hexane, 25/75, v/v
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.