Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Methyl Ethyl Ketone Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™
Click to view available options
Quantity:
3 x 1.2 mL
Description
Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. 2-Butanone (Methyl ethyl ketone, MEK), an aliphatic ketone, is considered as an aprotic and protophilic solvent. Its ability as an anticonvulsive agent in lithium-pilocarpine-induced status epilepticus in rats is reported.

Specifications
Specifications
CAS | 78-93-3 |
Ignition Point | 960°F |
Vapor Pressure | 71 mmHg (20°C) |
Vapor Density | 2.49 (vs. air) |
Density | 0.805 g/mL (at 25°C) |
Flash Point | -3°C |
Purity Grade Notes | These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available. |
Physical Form | Neat |
Linear Formula | C2H5 COCH3 |
MDL Number | MFCD00011648 |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction