Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Methyl Ethyl Ketone/Toluene, 90g/60g, Reagents
Click to view available options
Quantity:
4 L
Packaging:
Glass Bottle
Tin Can
Specifications
Specifications
| Color | Colorless |
| Quantity | 4 L |
| Packaging | Tin Can |
| Physical Form | Liquid |
| Specific Gravity | 0.8293 |
| Grade | Reagent |
| Chemical Name or Material | Methyl Ethyl Ketone/Toluene, 90 g/60 g |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction