Learn More
Methyl formate, 97%, may cont. up to ca 3% methanol
Description
Methyl formate is used as a precursor to formamide, dimethylformamide and formic acid. It is used as a blowing agent and utilized for quick-drying finishes due to its high vapor pressure. It is used as a component of azeotrope to recover expensive solvents. Further, it is used as a propellant and an intermediate for optical brighteners and hydrocyanic acid.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Methyl formate is used as a precursor to formamide, dimethylformamide and formic acid. It is used as a blowing agent and utilized for quick-drying finishes due to its high vapor pressure. It is used as a component of azeotrope to recover expensive solvents. Further, it is used as a propellant and an intermediate for optical brighteners and hydrocyanic acid.
Solubility
Miscible with water.
Notes
Incompatible with oxidizing agents.
Specifications
Specifications
| Melting Point | °C to 100°C |
| Density | 0.972 |
| Boiling Point | 32°C to 33°C |
| Flash Point | -26°C (-14°F) |
| Assay Percent Range | 97% |
| Refractive Index | 1.344 |
| Linear Formula | HCO2CH3 |
| Quantity | 500 mL |
| UN Number | UN1243 |
| Beilstein | 1734623 |
| Show More |
Safety and Handling
| Hazard Category | H224-H302-H315-H319-H331-H335 |
| Hazard Statement | GHS H Statement H224-H331-H371-H335-H302-H319 |
| Precautionary Statement | P210-P233-P240-P243-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P311-P330-P332+P313-P362-P372-P374-P380-P501c |
| DOTInformation | Transport Hazard Class: 3; Packing Group: I; Proper Shipping Name: METHYL FORMATE |
| EINECSNumber | 203-481-7 |
| RTECSNumber | LQ8925000 |
| TSCA | Yes |
| Recommended Storage | Keep cold |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.