Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl-Glutamine Rabbit anti-All, Polyclonal, MilliporeSigma™

Catalog No. ABS2185MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABS2185MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABS2185MI Supplier MilliporeSigma Supplier No. ABS2185
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-Methyl-Glutamine is a rabbit polyclonal antibody that detects glutamine methylated proteins. It has been tested for use in Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Methyl-Glutamine
Applications Western Blot
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Host Species Rabbit
Immunogen KLH-conjugated Gly-Gln (Me)-Gly tripeptide.
Purification Method Unpurified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Test Specificity This rabbit polyclonal antibody detects methylglutamine in proteins.
Target Species All species
Content And Storage Stable for 1 year at -20°C from date of receipt.Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.