Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Methyl Green 1%
SDP

Catalog No. 48003018 Shop All Novus Products
Change view
Click to view available options
Quantity:
50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
48-003-018 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 48-003-018 Supplier R&D Systems™ Supplier No. 48003018
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

R&D Systems™ Methyl Green 1% stain is free of crystal violet contamination and provides excellent contrast with DAB detection systems.

  • Uncontrolled product according to WHMIS classification criteria
  • There are no hazardous ingredients as defined in the United States 29 CFR 1910.1200 (OSHA) or the Canadian Controlled Products Regulations (WHMIS) and therefore not subject to the labelling requirements of the aforementioned standards

Specifications

Quantity 50 mL
Content And Storage Store the unopened product at room temperature
Product Line R&D Systems
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.