Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl Heptadecanoate 98.0+%, TCI America™
SDP

Catalog No. H056625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H056625G 25 g
H05665G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H056625G Supplier TCI America Supplier No. H056625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1731-92-6
Molecular Formula C18H36O2
Molecular Weight (g/mol) 284.484
MDL Number MFCD00009001
InChI Key HUEBIMLTDXKIPR-UHFFFAOYSA-N
Synonym methyl margarate, heptadecanoic acid, methyl ester, margaric acid methyl ester, heptadecanoic acid methyl ester, unii-41pzk7h0kl, 41pzk7h0kl, heptadecanoic acid,methyl ester, n-heptadecanoic acid methyl ester, methylheptadecanoate, formyl heptadecanoate
PubChem CID 15609
IUPAC Name methyl heptadecanoate
SMILES CCCCCCCCCCCCCCCCC(=O)OC

Specifications

Color White
Quantity 25 g
Formula Weight 284.48
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material Methyl Heptadecanoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.