Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl Heptafluorobutyrate 97.0+%, TCI America™
SDP

Catalog No. H103325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H103325G 25 g
H10335G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H103325G Supplier TCI America Supplier No. H103325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 356-24-1
Molecular Formula C5H3F7O2
Molecular Weight (g/mol) 228.066
MDL Number MFCD00000433
InChI Key MRPUVAKBXDBGJQ-UHFFFAOYSA-N
Synonym methyl heptafluorobutyrate, methyl heptafluorobutanoate, methyl perfluorobutyrate, methylheptafluorobutyrate, butanoic acid, heptafluoro-, methyl ester, heptafluorobutyric acid methyl ester, butyric acid, heptafluoro-, methyl ester, butanoic acid, 2,2,3,3,4,4,4-heptafluoro-, methyl ester, heptafluorobutyryl methyl ester, methyl perfluorbutanoate
PubChem CID 67741
IUPAC Name methyl 2,2,3,3,4,4,4-heptafluorobutanoate
SMILES COC(=O)C(C(C(F)(F)F)(F)F)(F)F

Specifications

Color Colorless
Boiling Point 81°C
Quantity 25 g
UN Number 3272
Formula Weight 228.07
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl Heptafluorobutyrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.