Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl hexanoate, 99%

Catalog No. AC449240010
Click to view available options
Quantity:
50 g
250 g
1 kg

CAS: 106-70-7 | C7H14O2 | 130.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106-70-7
Molecular Formula C7H14O2
Molecular Weight (g/mol) 130.19
MDL Number MFCD00009510
InChI Key NUKZAGXMHTUAFE-UHFFFAOYSA-N
Synonym methyl caproate, hexanoic acid methyl ester, hexanoic acid, methyl ester, methyl hexoate, methyl capronate, methyl n-hexanoate, methyl hexylate, caproic acid methyl ester, methyl caproate natural, methyl n-hexoate
PubChem CID 7824
ChEBI CHEBI:77322
IUPAC Name methyl hexanoate
SMILES CCCCCC(=O)OC

Specifications

Melting Point -71.0°C
Density 0.8840g/mL
Boiling Point 151.0°C
Flash Point 42°C
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4047 to 1.4067
Linear Formula CH3(CH2)4COOCH3
Quantity 1 kg
Specific Gravity 0.884
Solubility Information Solubility in water: insoluble
Viscosity <30 mPa.s (20°C)
Formula Weight 130.18
Percent Purity 99%
Chemical Name or Material Methyl hexanoate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Flammable liquid and vapor.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
EINECSNumber 203-425-1

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.