Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl-Histone H3 (Lys79) Mouse anti-Human, Clone: 3C7, Bioss
SDP

Catalog No. 501989948
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-9948 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-9948 Supplier Bioss Supplier No. BSM33098M
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Histones are relatively small proteins that contain a high proportion of positively charged amino acids, e. g., arginine and lysine. This positive charge allows histones to bind tightly to the negatively charged DNA. There are five basic types of histone molecules called H1, H2A, H2B, H3, and H4. Histones are abundantly present in virtually all eukaryotic cells.

Specifications

Antigen Methyl-Histone H3 (Lys79)
Applications Western Blot
Classification Monoclonal
Clone 3C7
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation TBS with 1% BSA, 50% glycerol and 0.03% Proclin 300; pH
Gene H3-4
Gene Accession No. Q16695
Gene Alias H3 histone family, member T; H3.4; H3/g; H3/t; H3FT; H3t; HIST3H3; histone 3, H3; histone cluster 3, H3; Histone H3.1t
Gene Symbols H3-4
Host Species Mouse
Immunogen KLH conjugated synthetic peptide derived from human Histone H3 (tri methyl K79).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8290
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.