Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl iso-Butyl Ketone, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
500mL
4L
Packaging:
Narrow Mouth Amber Glass Bottle

Chemical Identifiers

108-10-1
C6H12O
100.161
NTIZESTWPVYFNL-UHFFFAOYSA-N
7909
CHEBI:82344

High quality chemicals for laboratory and specialized industrial use

SMILES CC(C)CC(=O)C
IUPAC Name 4-methylpentan-2-one
Formula Weight 100.16
Physical Form Liquid
Packaging Narrow Mouth Amber Glass Bottle
Specific Gravity 1L = 0.80kg
Flash Point 14.00°C
Quantity 500mL
Chemical Name or Material Methyl iso-Butyl Ketone
Grade Baker Analyzed™ ACS Reagent, Meets ACS reagent chemical requirements