Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl iso-Butyl Ketone (Certified ACS), Fisher Chemical™

Catalog No. M213200 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
1 L
4 L
20 L
200 L
Packaging:
Glass Bottle
Steel Drum
Steel Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M213-200 200 L Steel Drum
M213-1 1 L Glass Bottle
M213-4 4 L Glass Bottle
M213-20 20 L Steel Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. M213-200
Add to Cart

Chemical Identifiers

CAS 108-10-1
Molecular Formula C6H12O
Molecular Weight (g/mol) 100.161
MDL Number MFCD00008938
InChI Key NTIZESTWPVYFNL-UHFFFAOYSA-N
Synonym 4-methyl-2-pentanone, methyl isobutyl ketone, isopropylacetone, isobutyl methyl ketone, hexone, mibk, 2-pentanone, 4-methyl, 2-methyl-4-pentanone, 4-methyl-2-oxopentane, methylisobutylketon
PubChem CID 7909
ChEBI CHEBI:82344
IUPAC Name 4-methylpentan-2-one
SMILES CC(C)CC(=O)C

Specifications

Melting Point -84°C
Density 0.80g/cm³
Quantity 200 L
Identification Pass Test
Packaging Steel Drum
Physical Form Liquid
Residue after Evaporation 0.005% max.
Assay Percent Range ≥98.5 %
Titratable Acid 0.002mEq/g max.
Vapor Pressure 19.9mmHg at 25°C
Water 0.1% max.
Boiling Point 117°C
Formula Weight 100.16
Percent Purity ≥98.5%
Grade ACS
Chemical Name or Material Methyl iso-Butyl Ketone
Show More Show Less
Safety handling Solvent Safety handling Check Alert storage code Red
Hazard Statement DANGER!
Precautionary Statement Emergency Overview
Flammable liquid and vapor. Harmful by inhalation. Irritating to eyes, respiratory system and skin. May cause central nervous system effects. Repeated exposure may cause skin dryness or cracking. Aspiration hazard if swallowed - can enter lungs and cause damage. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. If peroxide formation is suspected, do not open or move container. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes.

NFPA

Health:2
Flammability:3
Instability:1
DOTInformation DOT Class 3, : Flammable Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.